EMAIL THIS PAGE TO A FRIEND
Amyotrophic lateral sclerosis : official publication of the World Federation of Neurology Research Group on Motor Neuron Diseases
ALSUntangled 15: coconut Oil.
PMID 22471904