EMAIL THIS PAGE TO A FRIEND
Organic letters
Eight stereoisomers of homonojirimycin from D-mannose.
PMID 22472134
Abstract
Although there are 32 6-azidoheptitols, there are only 16 homonojirimycin (HNJ) stereoisomers. Two epimeric azidoalditols derived from d-mannose allow the synthesis in water of eight stereoisomers of HNJ.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
52388
α-Homonojirimycin, ≥98.0% (TLC)
C7H15NO5