EMAIL THIS PAGE TO A FRIEND
Annals of neurology
Implication of perturbed axoglial apparatus in early pediatric multiple sclerosis.
PMID 22473675
Abstract
Cerebrospinal fluid samples collected from children during initial presentation of central nervous system inflammation, who may or may not subsequently be diagnosed as having multiple sclerosis (MS), were subjected to large-scale proteomics screening. Unexpectedly, major compact myelin membrane proteins typically implicated in MS were not detected. However, multiple molecules that localize to the node of Ranvier and the surrounding axoglial apparatus membrane were implicated, indicating perturbed axon-glial interactions in those children destined for diagnosis of MS.