EMAIL THIS PAGE TO A FRIEND
Angewandte Chemie (International ed. in English)
Detergent-free formation and physicochemical characterization of nanosized lipid-polymer complexes: Lipodisq.
PMID 22473824