EMAIL THIS PAGE TO A FRIEND
Hepatology (Baltimore, Md.)
Serum autotaxin is increased in pruritus of cholestasis, but not of other origin, and responds to therapeutic interventions.
PMID 22473838
Abstract
Pruritus is a seriously disabling symptom accompanying many cholestatic liver disorders. Recent experimental evidence implicated the lysophospholipase, autotaxin (ATX), and its product, lysophosphatidic acid (LPA), as potential mediators of cholestatic pruritus. In this study, we highlight that increased serum ATX levels are specific for pruritus of cholestasis, but not pruritus of uremia, Hodgkin's disease, or atopic dermatitis. Treatment of patients with cholestasis with the bile salt sequestrant, colesevelam, but not placebo, effectively reduced total serum bile salts and fibroblast growth factor 19 levels, but only marginally altered pruritus intensity and ATX activity. Rifampicin (RMP) significantly reduced itch intensity and ATX activity in patients with pruritus not responding to bile salt sequestrants. In vitro, RMP inhibited ATX expression in human HepG2 hepatoma cells and hepatoma cells overexpressing the pregnane X receptor (PXR), but not in hepatoma cells in which PXR was knocked down. Treatment of severe, refractory pruritus by the molecular adsorbents recirculation system or nasobiliary drainage improved itch intensity, which, again, correlated with the reduction of ATX levels. Upon reoccurrence of pruritus, ATX activity returned to pretreatment values. Serum ATX activity is specifically increased in patients with cholestatic, but not other forms of, systemic pruritus and closely correlates with the effectiveness of therapeutic interventions. The beneficial antipruritic action of RMP may be explained, at least partly, by the PXR-dependent transcriptional inhibition of ATX expression. Thus, ATX likely represents a novel therapeutic target for pruritus of cholestasis.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
241075
Allylamine, ≥99%
C3H7N
145831
Allylamine, 98%
C3H7N