EMAIL THIS PAGE TO A FRIEND
Organic letters
TiCl4-promoted tandem carbonyl or imine addition and Friedel-Crafts cyclization: synthesis of benzo-fused oxabicyclooctanes and nonanes.
PMID 22475177
Abstract
A new and convenient synthesis of benzo-fused 8-oxabicyclo[3.2.1]octane and 9-oxabicyclo[4.2.1]nonane derivatives are described. The reaction involved a TiCl(4)-mediated tandem carbonyl or imine addition followed by a Friedel-Crafts cyclization to provide these functionalized derivatives in good to excellent yields and high diastereoselectivity.