EMAIL THIS PAGE TO A FRIEND
Bulletin of environmental contamination and toxicology
Estimation of etofenprox residues in tomato fruits by QuEChERS methodology and HPLC-DAD.
PMID 22476259
Abstract
Etofenprox residues were estimated by employing standardized QuEChERS technique in tomato following one application of Trebon(®) 20 % EC. The average recoveries of etofenprox on tomato for fortification levels 0.01, 0.1 and 0.5 mg/kg were observed to be 87.5 %, 89.7 % and 92.2 %, respectively, with relative standard division of 3.50, 4.11 and 3.20. The LOQ for tomato was found to be 0.01 mg/kg. The average initial deposit of etofenprox on tomato was observed to be 0.783 mg/kg, at single application rate. This etofenprox residue dissipated below its LOQ of 0.01 after 15 days at a single dosage. Half-life of etofenprox was observed to be 2.15 days, at the recommended dosage. These data could provide guidance for the proper and safe use of this pesticide on tomato in Egypt.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
34094
Etofenprox, PESTANAL®, analytical standard
C25H28O3