PloS one
Granzyme B cleaves decorin, biglycan and soluble betaglycan, releasing active transforming growth factor-β1.
PMID 22479366
Abstract
Granzyme B (GrB) is a pro-apoptotic serine protease that contributes to immune-mediated target cell apoptosis. However, during inflammation, GrB accumulates in the extracellular space, retains its activity, and is capable of cleaving extracellular matrix (ECM) proteins. Recent studies have implicated a pathogenic extracellular role for GrB in cardiovascular disease, yet the pathophysiological consequences of extracellular GrB activity remain largely unknown. The objective of this study was to identify proteoglycan (PG) substrates of GrB and examine the ability of GrB to release PG-sequestered TGF-β1 into the extracellular milieu. Three extracellular GrB PG substrates were identified; decorin, biglycan and betaglycan. As all of these PGs sequester active TGF-β1, cytokine release assays were conducted to establish if GrB-mediated PG cleavage induced TGF-β1 release. Our data confirmed that GrB liberated TGF-β1 from all three substrates as well as from endogenous ECM and this process was inhibited by the GrB inhibitor 3,4-dichloroisocoumarin. The released TGF-β1 retained its activity as indicated by the induction of SMAD-3 phosphorylation in human coronary artery smooth muscle cells. In addition to contributing to ECM degradation and the loss of tissue structural integrity in vivo, increased extracellular GrB activity is also capable of inducing the release of active TGF-β1 from PGs.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D7910
3,4-Dichloroisocoumarin, serine protease inhibitor
C9H4Cl2O2