EMAIL THIS PAGE TO A FRIEND
Organic letters
Lewis acid promoted highly diastereoselective Petasis Borono-Mannich reaction: efficient synthesis of optically active β,γ-unsaturated α-amino acids.
PMID 22480132
Abstract
An efficient and straightforward method for the preparation of highly enantiomerically enriched β,γ-unsaturated α-amino acid derivatives by a Lewis acid promoted diastereoselective Petasis reaction of vinylboronic acid, N-tert-butanesulfinamide, and glyoxylic acid has been developed. The synthetic utilities of the approach were demonstrated by the rapid and convenient construction of challenging cyclopenta[c]proline derivatives.
Related Materials