American journal of kidney diseases : the official journal of the National Kidney Foundation
Hepatorenal syndrome: a severe, but treatable, cause of kidney failure in cirrhosis.
PMID 22480795
Hepatorenal syndrome (HRS) is a unique type of kidney failure that occurs in advanced cirrhosis. It is characterized by functional impairment of the kidneys due to vasoconstriction of the renal arteries in the setting of preserved tubular function and absence of significant histologic abnormalities. Renal vasoconstriction in HRS is due to severe vasodilation of the splanchnic arteries associated with portal hypertension, leading to a decrease in effective arterial blood volume and arterial pressure. HRS commonly develops after a trigger, usually a bacterial infection, that disrupts the arterial circulation, but it also may occur spontaneously. There are 2 forms of HRS: type 1 is characterized by an acute progressive decrease in kidney function and very short survival without treatment, whereas type 2 features stable less severe kidney failure and longer survival compared with type 1. A liver transplant is the preferred treatment for HRS. Pharmacologic treatment with vasoconstrictors to reverse splanchnic vasodilation, together with albumin, is effective in 40%-50% of patients with type 1 HRS and improves survival. The drug of choice is the vasopressin analogue terlipressin. Renal replacement therapy should not be used as first-line therapy.
Related Materials
Product #
Molecular Formula
Add to Cart
[Lys8]-Vasopressin, ≥98% (HPLC)