EMAIL THIS PAGE TO A FRIEND
Annals of hepatology
Living Fasciola hepatica in biliary tree: a case report.
PMID 22481460
Abstract
Here we report a rare case of living Fasciola hepatica in biliary tract. The patient was in acute phase of infection and treated successfully with 10 mg/kg oral triclabendazole after the fluke was extracted using endoscopic retrograde cholangiopancreatography (ERCP).
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
32802
Triclabendazole, VETRANAL, analytical standard
C14H9Cl3N2OS