EMAIL THIS PAGE TO A FRIEND
Dentistry today
Integrating techniques to restore an adolescent's smile.
PMID 22482204
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
14372 Pumice stone, granulated