EMAIL THIS PAGE TO A FRIEND
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy
SERS and DFT investigation of 1-(2-pyridylazo)-2-naphthol and its metal complexes with Al(III), Mn(II), Fe(III), Cu(II), Zn(II) and Pb(II).
PMID 22484262
Abstract
The development of surface-enhanced Raman scattering (SERS) as a prospective analytical methodology for detection of metal ions was shown in recent years by several studies on metal complexes. In this work, 1-(2-pyridylazo)-2-naphthol (PAN) and its Al(III), Mn(II), Fe(III), Cu(II), Zn(II) and Pb(II) complexes were studied by FTIR, FT-Raman and surface enhanced Raman spectroscopies. Molecular geometry optimization, molecular electrostatic potential (MEP) distribution and vibrational frequencies calculations were performed using the hybrid B3LYP exchange-correlation functional for the PAN molecule and its bidentate complexes. The calculated MEP distributions indicated the atoms with highest electronegativity, the adsorption to the silver surface occurring through these atoms. Based on experimental and theoretical data we were able to identify unique and representative features, useful for the identification of each PAN-metal complex.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
101036
1-(2-Pyridylazo)-2-naphthol, indicator grade
C15H11N3O