EMAIL THIS PAGE TO A FRIEND
Bulletin of environmental contamination and toxicology
Tolerance and bioaccumulation of copper by the entomopathogen Beauveria bassiana (Bals.-Criv.) Vuill. exposed to various copper-based fungicides.
PMID 22484327
Abstract
This work evaluates for the first time the relationships between copper-tolerance, -solubilization and -bioaccumulation in the entomopathogen Beauveria bassiana exposed to Bordeaux mixture, copper oxychloride or copper hydroxide. Bordeaux mixture was highly detrimental to fungus, by inhibiting the growth totally at the recommended dose (RD) and 2×RD. Copper hydroxide and copper oxychloride were found to be less toxic, reducing fungus growth, sporulation and conidial germination in an average of 29  %, 30 % and 58 %, respectively. These two copper forms were the easiest to solubilize, to precipitate and the most accumulated by B. bassiana, suggesting the involvement of all these processes on fungus copper-tolerance.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
N11502 Copper oxychloride, technical grade