EMAIL THIS PAGE TO A FRIEND
Bioscience, biotechnology, and biochemistry
Properties of Jack bean α-mannosidase in the presence of hyaluronan.
PMID 22484962
Abstract
An enzymatic reaction within a mesh-like structure constructed using hyaluronan was investigated in order to understand the influence of specific reaction environments in a living body on the reaction. This mesh-like structure, which mimicked extracellular matrix conditions, was found to accelerate glycohydrolysis by Jack bean α-mannosidase.