Neurochemical research
Abnormal gangliosides are localized in lipid rafts in Sanfilippo (MPS3a) mouse brain.
PMID 22484966
Allogenic stem cell transplantation can reduce lysosomal storage of heparan sulfate-derived oligosaccharides by up to 27 % in Sanfilippo MPS3a brain, but does not reduce the abnormal storage of sialolactosylceramide (G(M3)) or improve neurological symptoms, suggesting that ganglioside storage is in a non-lysosomal compartment. To investigate this further we isolated the Triton X100-insoluble at 4 °C, lipid raft (LR) fraction from a sucrose-density gradient from cerebral hemispheres of a 7 month old mouse model of Sanfilippo MPS3a and age-matched control mouse brain. HPLC/MS/MS analysis revealed the expected enrichment of normal complex gangliosides, ceramides, galatosylceramides and sphingomyelin enrichment in this LR fraction. The abnormal HS-derived oligosaccharide storage material was in the Triton X100-soluble at 4 °C fractions (8-12),whereas both GM3 and sialo[GalNAc]lactosylceramide (GM2) were found exclusively in the LR fraction (fractions 3 and 4) and were >90 % C18:0 fatty acid, suggesting a neuronal origin. Further analysis also revealed a >threefold increase in the late-endosome marker bis (monoacylglycerol) phosphate (>70 % as C22:6/22:6-BMP) in non-LR fractions 8-12 whereas different forms of the proposed BMP precursor, phosphatidylglycerol (PG) were in both LR and non-LR fractions and were less elevated in MPS3a brain. Thus heparan sulfate-derived oligosaccharide storage is associated with abnormal lipid accumulation in both lysosomal (BMP) and non-lysosomal (GM3 and GM2) compartments.
Related Materials
Product #
Molecular Formula
Add to Cart
G8397 Monosialoganglioside GM2 from bovine brain, ≥95% (TLC)