EMAIL THIS PAGE TO A FRIEND
Zeitschrift fur Naturforschung. C, Journal of biosciences
Evaluation of the anti-inflammatory and antinociceptive properties of p-cymene in mice.
PMID 22486037
Abstract
We attempted to identify the antinociceptive and anti-inflammatory actions of the monoterpene p-cymene. Firstly, behavioural screening was carried out to verify the influence of p-cymene [25, 50, and 100 mg/kg intraperitoneal (i.p.)] on the central nervous system (CNS) activity. The antinociceptive activity of p-cymene was evaluated by the acetic acid-induced writhing response, formalin, and hot-plate test, respectively. The leukocyte migration induced by injection of carrageenan was used to assess the anti-inflammatory activity. p-Cymene showed depressant activity on CNS after 4 h of treatment and also a possible action on the autonomous nervous system (ANS), mainly at the dose of 100 mg/kg (i.p.). It was found that p-cymene (50 and 100 mg/kg, i.p.) significantly (p < 0.05) reduced the writhing responses induced by acetic acid. p-Cymene also decreased the licking time in the first and second phase, respectively, of the formalin test. The results of the hot-plate test showed that all doses of p-cymene increased significantly the latency time of the response to the thermal stimulus in both licking and jumping parameters. In addition, there was a significantly (p < 0.05) decreased leukocyte migration at all doses of p-cymene. The experimental data demonstrate that p-cymene possesses antinociceptive and anti-inflammatory activities.