EMAIL THIS PAGE TO A FRIEND
Chemical communications (Cambridge, England)
Direct asymmetric Mannich reaction of phthalides: facile access to chiral substituted isoquinolines and isoquinolinones.
PMID 22488217
Abstract
The first Mannich reaction employing phthalides using a quinidine-based multifunctional catalyst has been developed. The reported method led to the synthesis of 3,3-disubstituted phthalide derivatives in excellent yields, with good diastereo- and enantioselectivities. Convenient synthesis of chiral isoquinolinones and isoquinolines has also been demonstrated.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P39605
Phthalide, 98%
C8H6O2