EMAIL THIS PAGE TO A FRIEND
Chemistry, an Asian journal
Synthesis of optically pure norcantharidin analogue NCA-01, a highly selective protein phosphatase 2B inhibitor, and its derivatives.
PMID 22488808
Abstract
An efficient synthetic route to optically pure norcantharidin analogue NCA-01, a highly selective inhibitor of protein phosphatase 2B (PP2B; calcineurin), has been developed. The absolute stereochemistry of the enantiomers was determined by X-ray crystallographic analysis. Optically pure NCA derivatives that had various substituents at the C1 position were synthesized in a similar manner. The PP2B-inhibitory activities of NCA-01 and its derivatives were independent of the enantiomeric form. NCA-01 dimethyl ester potently inhibited IL-2 production in Jurkat cells.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
N8784
Norcantharidin, solid
C8H8O4