EMAIL THIS PAGE TO A FRIEND
ChemMedChem
Selective modification of the N-terminal structure of polytheonamide B significantly changes its cytotoxicity and activity as an ion channel.
PMID 22489077
Abstract
Chemical point mutation: Polytheonamide B is a naturally occurring polypeptide containing 48 amino acids. It both displays potent cytotoxicity and acts as a monovalent cation channel in vitro. Chemoselective methods to modify the 44th, N-, and C-terminal residues of the natural product have been developed, and evaluation of the resultant derivatives suggests that the intrinsic activities of the peptide can only be altered by switching its N-terminal substitution.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
08010
Safrole, certified reference material, TraceCERT®
C10H10O2
S9652
Safrole, ≥97%
C10H10O2