EMAIL THIS PAGE TO A FRIEND
Langmuir : the ACS journal of surfaces and colloids
Oil-in-alcohol highly concentrated emulsions as templates for the preparation of macroporous materials.
PMID 22489569
Abstract
New oil-in-alcohol highly concentrated emulsions were formulated and were used as a templates to obtain macroporous poly(furfuryl alcohol) monoliths by a one-step method. The oil-in-alcohol highly concentrated emulsions were prepared by stepwise addition of the oil phase to the surfactant-alcohol solution and were characterized by optical microscopy and by laser diffraction. The typical structure of highly concentrated emulsions, with close-packed polyhedral droplets, has been observed. Poly(furfuryl alcohol) monoliths were obtained by polymerizing in the external phase of these emulsions. These materials are mainly macroporous and retain the size distribution and morphology from the highly concentrated emulsions. The internal structure of the monoliths was observed by scanning electron microscopy. The images showed an interconnected network with pore size similar to the droplet size of the highly concentrated emulsions used as templates.
Related Materials