EMAIL THIS PAGE TO A FRIEND
Journal of applied physiology (Bethesda, Md. : 1985)
Effects of β₂-agonists on force during and following anoxia in rat extensor digitorum longus muscle.
PMID 22492937
Abstract
Electrical stimulation of isolated muscles may lead to membrane depolarization, gain of Na(+), loss of K(+) and fatigue. These effects can be counteracted with β(2)-agonists possibly via activation of the Na(+)-K(+) pumps. Anoxia induces loss of force; however, it is not known whether β(2)-agonists affect force and ion homeostasis in anoxic muscles. In the present study isolated rat extensor digitorum longus (EDL) muscles exposed to anoxia showed a considerable loss of force, which was markedly reduced by the β(2)-agonists salbutamol (10(-6) M) and terbutaline (10(-6) M). Intermittent stimulation (15-30 min) clearly increased loss of force during anoxia and reduced force recovery during reoxygenation. The β(2)-agonists salbutamol (10(-7)-10(-5) M) and salmeterol (10(-6) M) improved force development during anoxia (25%) and force recovery during reoxygenation (55-262%). The effects of salbutamol on force recovery were prevented by blocking the Na(+)-K(+) pumps with ouabain or by blocking glycolysis with 2-deoxyglucose. Dibutyryl cAMP (1 mM) or theophylline (1 mM) also improved force recovery remarkably. In anoxic muscles, salbutamol decreased intracellular Na(+) and increased (86)Rb uptake and K(+) content, indicating stimulation of the Na(+)-K(+) pumps. In fatigued muscles salbutamol induced recovery of excitability. Thus β(2)-agonists reduce the anoxia-induced loss of force, leading to partial force recovery. These data strongly suggest that this effect is mediated by cAMP stimulation of the Na(+)-K(+) pumps and that it is not related to recovery of energy status (PCr, ATP, lactate).