EMAIL THIS PAGE TO A FRIEND
Circulation
Cardiac amyloidosis mimicking hypertrophic cardiomyopathy with obstruction: treatment with disopyramide.
PMID 22492950
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D6035 Disopyramide phosphate salt
C21H29N3O · H3O4P