EMAIL THIS PAGE TO A FRIEND
Journal of environmental science and health. Part. B, Pesticides, food contaminants, and agricultural wastes
Effects of the organic matter from swine wastewater on the adsorption and desorption of alachlor in soil.
PMID 22494371
Abstract
The application of swine wastewater to the soil for agricultural purposes results in the addition of total and dissolved organic matter to the soil, which may interfere with the dynamics of pesticides in the soil. The objective of this study was to evaluate the effects of the application of total and dissolved organic matter from a biodigester and a treatment lagoon of swine wastewater in the adsorption and desorption of alachlor [2-chloro-2,6-diethyl-N(methoxymethyl acetamide)]. The assay was performed by the batch equilibrium method, and the results were fitted to the Freundlich model. The curve comparison test revealed a greater adsorption of alachlor in the soil treated with swine wastewater from the biodigester. The adsorption and desorption of alachlor increased in the soils where swine wastewater was added, and hysteresis was observed in all of the treatments.
Related Materials