EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry
Design and synthesis of 4-O-methylhonokiol analogs as inhibitors of cyclooxygenase-2 (COX-2) and PGF₁ production.
PMID 22494844
Abstract
A series of novel 4-O-methylhonokiol analogs were synthesized in light of revealing structure-activity relationship for inhibitory effect of COX-2 enzyme. The key strategy of the molecular design was oriented towards modification of the potential metabolic soft spots (e.g., phenol and olefin) or by altering the polar surface area via incorporating heterocycles such as isoxazole and triazole. Most of all exhibited the inhibitory effects on COX-2 and PGF(1) production but not macrophage NO production. Especially, aryl carbamates 10 and 11 exhibited more potent inhibitory activity against COX-2 and PGF(1) production.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P5765
Prostaglandin F, synthetic
C20H36O5