EMAIL THIS PAGE TO A FRIEND
Journal of hypertension
Clarifying the role of preoperative α-blockade in pheochromocytoma?
PMID 22495141
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D9815
Doxazosin mesylate, ≥97% (HPLC), powder
C23H25N5O5 · CH3SO3H