EMAIL THIS PAGE TO A FRIEND
Bioorganicheskaia khimiia
[Synthesis of 2'-aminometylmorpholino nucleoside analogues containing 4'-carboxymethyl lynker group].
PMID 22497082
Abstract
A simple and effective method for the synthesis of 2'-aminomethylmorpholino-4'-carboxymethyl nucleoside analogues and Boc-modified derivatives as synthons for peptide synthesis was developed.