EMAIL THIS PAGE TO A FRIEND
Organic letters
Dithiafulvenyl unit as a new donor for high-efficiency dye-sensitized solar cells: synthesis and demonstration of a family of metal-free organic sensitizers.
PMID 22497498
Abstract
This work identifies the dithiafulvenyl unit as an excellent electron donor for constructing D-π-A-type metal-free organic sensitizers of dye-sensitized solar cells (DSCs). Synthesized and tested are three sensitizers all with this donor and a cyanoacrylic acid acceptor but differing in the phenyl (DTF-C1), biphenyl (DTF-C2), and phenyl-thiopheneyl-phenyl π-bridges (DTF-C3). Devices based on these dyes exhibit a dramatically improved performance with the increasing π-bridge length, culminating with DTF-C3 in η = 8.29% under standard global AM 1.5 illumination.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
431966
4-Formylphenylboronic acid, ≥95.0%
C7H7BO3