EMAIL THIS PAGE TO A FRIEND
Veterinary ophthalmology
Electroretinogram responses of the normal thoroughbred horse sedated with detomidine hydrochloride.
PMID 22498137
Abstract
The main objective was to record electroretinogram (ERG) parameters of normal thoroughbred mares using the HMsERG, a mini-Ganzfeld electroretinographic unit, and a contact lens electrode. The second objective was to determine whether IV detomidine hydrochloride at 0.015 mg/kg is consistently an effective choice for sedation of horses undergoing this ERG protocol. The study population consisted of 30 normal thoroughbred mares. ERG data were harvested using a protocol that included three different light intensities (10, 3000, and 10,000 mcd s/m(2)) and a 30-Hz flicker at 3000 mcd s/m(2). Mean, median, standard deviation, and estimated normal ranges using the 5-95% of the data for a- and b-wave implicit times (IT), amplitudes (AMP), and b/a ratios were reported. Scotopic results at low intensity (10 mcd s/m(2)) had estimated ranges for b-wave IT of 41.8-72.9 ms and AMP of 19.8-173.3 μV. Middle intensity (3000 mcd s/m(2)) a-wave IT was 13.2-14.7 ms with a-wave AMP of 68.4-144 μV; the b-wave IT was 28.7-41.5 ms with b-wave AMP of 105.7-271.5 μV; and the b/a ratio was 0.95-2.71. The high-intensity (10,000 mcd s/m(2)) average recordings showed an a-wave IT of 13-14.9 ms, a-wave AMP of 85.7-186.8 μV; b-wave IT of 26.6-45.4 ms, b-wave AMP of 104.7-250.6 μV; and a b/a wave ratio of 0.7-2.0. The 30-Hz cone flicker showed an IT of 22.8-28.9 ms and AMP of 44.1-117.1 μV. Results of normal thoroughbred ERG responses are reported. The protocol proved to be simple and safe and provided consistent results.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
Y0000001 Detomidine hydrochloride, European Pharmacopoeia (EP) Reference Standard
C12H14N2 · HCl