EMAIL THIS PAGE TO A FRIEND
Chemical communications (Cambridge, England)
Novel D-π-A structured Zn(II)-porphyrin dyes containing a bis(3,3-dimethylfluorenyl)amine moiety for dye-sensitised solar cells.
PMID 22499080
Abstract
A high solar-to-electricity conversion efficiency of 7.22% was achieved with a short circuit current (J(sc)) of 15.30 mA cm(-2), an open circuit voltage (V(oc)) of 669 mV and a fill factor (FF) of 0.71 for the 2Flu-ZnP-CN-COOH dye with a multi-functional co-adsorbent, under 100 mW cm(-2) AM 1.5 G simulated light.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
900387
Bis(9,9-dimethyl-9H-fluoren-7-yl)amine
C30H27N