EMAIL THIS PAGE TO A FRIEND
Poultry science
Characterization of 5-hydroxytryptamine-induced contraction and acetylcholine-induced relaxation in isolated chicken basilar artery.
PMID 22499874
Abstract
The aim of the present study was to clarify the responsiveness of the chicken basilar artery to 5-hydroxytryptamine (5-HT) and acetylcholine (ACh) and to characterize the related receptor subtypes in vitro. Basilar arteries were obtained from freshly slaughtered broiler chickens. The 5-HT induced concentration-dependent contraction of the arteries. The concentration-response curves for 5-HT were shifted 30-fold to the right by methiothepin (a 5-HT(1) and 5-HT(2) receptor antagonist) and 3-fold to the right by ketanserin (a 5-HT(2) receptor antagonist). In the presence of ketanserin, the concentration-response curve for 5-HT was shifted 10-fold to the right by methiothepin. The pA(2) value for methiothepin was 8.26. The ACh induced concentration-dependent relaxation under conditions of precontraction by 5-HT. The concentration-response curve for ACh was shifted to the right by atropine [a nonselective muscarinic (M) receptor antagonist] and hexahydro-sila-difenidol hydrochloride, a p-fluoroanalog (pFHHSiD, an M(3) receptor antagonist), but not by pirenzepine (an M(1) receptor antagonist) or methoctramine (an M(2) receptor antagonist). The pA(2) value for pFHHSiD was 7.55. Nω-Nitro-l-arginine (a nitric oxide synthase inhibitor) inhibited ACh-induced relaxation by approximately 50%. These results suggest that 5-HT induces contraction via activation of 5-HT(1) and 5-HT(2) receptors and that ACh induces relaxation via activation of the M(3) receptor. The 5-HT(1) receptor might play a dominant role in 5-HT-induced contraction. One of the factors involved in ACh-induced relaxation is probably nitric oxide released from endothelial cells.