EMAIL THIS PAGE TO A FRIEND
Sexually transmitted diseases
Clinical burden of herpes simplex virus disease in people with human immunodeficiency virus.
PMID 22504602
Abstract
Serologic studies indicate that herpes simplex virus (HSV)-1 and HSV-2 infections are highly prevalent among people infected with HIV. As an ulcerative genital disease, HSV may be important to HIV transmission and HIV-comorbidity. Routine clinical care of HSV in this population has not been described. Data were abstracted from medical records of HIV-infected individuals by the Adult/Adolescent Spectrum of HIV Disease Project. Clinician-documented HSV diagnosis and HSV treatment, defined as any prescription for acyclovir, valacyclovir, or famciclovir, were the outcomes of interest. We present descriptive statistics and trends in HSV diagnosis and treatment. Between 1989 and 2004, 61,299 people were followed in this study. HSV was diagnosed in 20% of the population, and 32% of the population received HSV antiviral prescriptions. Prescriptions for episodic treatment were given to 28% of patients, and 11% received prescriptions for suppressive therapy. The average annual rate of HSV diagnosis declined by 31% during the course of the study. Clinically recognized HSV infections were frequent despite declining rates of diagnosis. Providers should have a high index of suspicion for HSV and consider routine screening and suppressive therapy for patients at risk of clinical disease.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
F7932
Famciclovir, ≥98% (HPLC)
C14H19N5O4