EMAIL THIS PAGE TO A FRIEND
Advanced materials (Deerfield Beach, Fla.)
Understanding charge transfer at PbS-decorated graphene surfaces toward a tunable photosensor.
PMID 22505476
Abstract
An intrinsic mechanism of photoinduced hole transfer reactions occurring at the grapheme-PbS interface is described with the purpose of building a tunable photosensor with a responsivity of more than 10(3) A W(-1) . It is remarkable that rational utilization of this finding also realizes symmetric, opposing photoswitching effects, which are effectively mirror images, in a single pristine graphene device. These results highlight the vital importance of interface modification as a powerful tool for creating future ultrasensitive optoelectronic devices.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
372595
Lead(II) sulfide, 99.9% trace metals basis
PbS