EMAIL THIS PAGE TO A FRIEND
La Medicina del lavoro
[A method for measuring urinary concentrations of benzene. Its use in monitoring of subjects exposed to low levels].
PMID 2250606
Abstract
Benzene is a widely diffuse solvent (atmosphere, cigarette smoke, some foods); in the industrial environment benzene is currently present at concentrations of ppm. A valid method of biological monitoring that is easy to perform is needed for assessing occupational and non-occupational exposures. A new method has been developed to evaluate low concentrations of benzene in urine samples by means of a "dynamic" headspace (50 ml of urine in a 120 ml vial). The urine is saturated with anhydrous Na2SO4 in order to support the entrance of benzene in the air over the urine. The solvent is stripped from the urine surface and concentrated on an adsorbent substrate (Carbotrap 100 tube) by means of a suction pump (150 ml/min). A simultaneous intake of filtered air through a charcoal tube allows wash-up of the headspace. Benzene is thermically desorbed and injected in a column (Thermal tube desorber-Supelco; 370 degrees C thermal flash; borosilicate capillary glass column SPB-1 60 m length, 0.75 mm I.D., 1 micron film thickness; G.C. Dani 8580-FID). The detection limit of the method is about 50 ng/l and the variation coefficient is 4.7%. The method was checked on urine samples of 5 non-smokers and 5 smokers: mean values of 135 and 944 ng/l respectively were obtained. A further analysis on urine samples of 60 smokers revealed a significant relationship (p less than 0.001) between urinary benzene concentrations and C0 alveolar concentrations (r = 0.626). A close relationship between benzene exposure levels and urinary concentrations was found in a group of workers exposed to low environmental benzene concentrations (mean value 1200 micrograms/m3) (r = 0.763).
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
25087
Carbotrap® 100, glass TD tube, preconditioned
20242 Carbotrap® 200 Dynatherm 890, 850, for use with Dynatherm ACEM-9300, ACEM-900 850/890 TDU, MTDU and OI Analytical Air Tube Desorber or DMP-16
20865 Carbotrap® 201, for use with Dynatherm ACEM 900/901-FF, I.D. × O.D. × L 1 mm × 6 mm × 7.25 in.
20083-U
Carbotrap® 202, stainless steel TD tube, O.D. × L 1/4  in. × 3 1/2  in., preconditioned, pkg of 10 ea
29745-U
Carbotrap® 202, O.D. × L 1/4  in. × 3 1/2  in., stainless steel TD tube, Sealed with Brass Endcaps, preconditioned, pkg of 10 ea
20013-U
Carbotrap® 217, stainless steel TD tube, O.D. × L 1/4  in. × 3 1/2  in., preconditioned, pkg of 10 ea
29531-U Carbotrap® 217, glass TD tube, fritted, O.D. × L 1/4  in. × 3 1/2  in., preconditioned, pkg of 10 ea
30138-U Carbotrap® 217, glass TD tube, fritted, O.D. × L 1/4  in. × 3 1/2 , unconditioned, pkg of 10 ea
30137-U
Carbotrap® 217, stainless steel TD tube, O.D. × L 1/4 × 3 1/2 , unconditioned, pkg of 10 ea
29744-U
Carbotrap® 217, O.D. × L 1/4  in. × 3 1/2  in., stainless steel TD tube, Sealed with Brass Endcaps, preconditioned, pkg of 10 ea
29750-U Carbotrap® 217, O.D. × L 1/4  in. × 3 1/2  in., glass TD tube, fritted, Sealed with Brass Endcaps, preconditioned, pkg of 10 ea
20875
Carbotrap® 300, glass TD tube, fritted, O.D. × I.D. × L 6 mm × 4 mm × 4.5 in., preconditioned, pkg of 1 ea
28283-U
Carbotrap® 300, glass TD tube, fritted, O.D. × I.D. × L 6 mm × 4 mm × 7 in., preconditioned, pkg of 1 ea
28273-U
Carbotrap® 300, stainless steel TD tube, O.D. × I.D. × L 6 mm × 4 mm × 7 in., preconditioned, pkg of 1 ea
20370-U Carbotrap® 300, stainless steel TD tube, O.D. × I.D. × L 1/4  in. × 5 mm × 7 in., preconditioned, pkg of 1 ea
21705-U
Carbotrap® 300, O.D. × L 1/4  in. × 3 1/2  in., stainless steel TD tube, preconditioned, pkg of 10 ea
29532-U
Carbotrap® 300, glass TD tube, fritted, O.D. × L 1/4  in. × 3 1/2  in., preconditioned, pkg of 10 ea
30141-U
Carbotrap® 300, glass TD tube, fritted, O.D. × L 1/4  in. × 3 1/2  in., unconditioned, pkg of 10 ea
30139-U
Carbotrap® 300, stainless steel TD tube, unconditioned
29746-U
Carbotrap® 300, O.D. × L 1/4  in. × 3 1/2  in., stainless steel TD tube, Sealed with Brass Endcaps, preconditioned, pkg of 10 ea
29751-U
Carbotrap® 300, O.D. × L 1/4  in. × 3 1/2  in., glass TD tube, fritted, Sealed with Brass Endcaps, preconditioned, pkg of 10 ea
20881 Carbotrap® 317, glass TD tube (fast-flow), preconditioned
28311-U Carbotrap® 349, glass TD tube, fritted, O.D. × I.D. × L 6 mm × 4 mm × 7 in., preconditioned, pkg of 1 ea
29752-U Carbotrap® 349, O.D. × L 1/4  in. × 3 1/2  in., glass TD tube, fritted, Sealed with Brass Endcaps, preconditioned, pkg of 10 ea
20287 Carbotrap® Adsorbent, matrix Carbotrap® B, 20-40 mesh, bottle of 10 g
11325-U Carbotrap® Adsorbent, matrix Carbotrap® B, 20-40 mesh, pack of 144 × 190 mg
20309 Carbotrap® Adsorbent, matrix Carbotrap® C, 20-40 mesh, bottle of 10 g
11047-U Carbotrap® Adsorbent, matrix Carbotrap® C, 20-40 mesh, bottle of 500 g
10435-U Carbotrap® Adsorbent, matrix Carbotrap® X, 20-40 mesh, bottle of 10 g
10460-U Carbotrap® Adsorbent, matrix Carbotrap® Y, 20-40 mesh, bottle of 10 g