EMAIL THIS PAGE TO A FRIEND
Organic letters
Scandium(III)-catalyzed enantioselective allylation of isatins using allylsilanes.
PMID 22506841
Abstract
The scandium(III)-catalyzed enantioselective Hosomi-Sakurai allylation of isatins with various substituted allylic silanes is described. A catalyst loading as low as 0.05 mol % is utilized at room temperature to afford the 3-allyl-3-hydroxy-2-oxindoles in excellent yields and enantioselectivity up to 99% ee, including a demonstration of a gram-scale reaction. The effects of additives and varying silyl groups were explored to demonstrate the scope and application.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
261262
Scandium, powder, 99.9% trace rare earth metals basis
Sc