EMAIL THIS PAGE TO A FRIEND
Biochemical and biophysical research communications
The tyrosine phosphatase HD-PTP (PTPN23) is degraded by calpains in a calcium-dependent manner.
PMID 22510412
Abstract
HD-PTP (PTPN23) is a non-transmembrane protein tyrosine phosphatase which contributes to the signal transduction pathways involved in the regulation of cell migration and invasion. We here demonstrate in T24 bladder carcinoma cells that HD-PTP undergoes calcium-dependent degradation which can be prevented by specific calpain inhibitors. In addition, treatment of the cells with the calpain inhibitor calpeptin results in the redistribution of endogenous HD-PTP to the periphery of the cells. Since (i) calpains are overexpressed in some tumors and (ii) the downregulation of HD-PTP enhances cell migration and invasion, we propose that HD-PTP degradation by calpains might result in the acquisition of a more aggressive phenotype in neoplastic cells.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C8999
Calpeptin, ≥98% (HPLC)
C20H30N2O4