EMAIL THIS PAGE TO A FRIEND
Chemical communications (Cambridge, England)
Facile synthesis of graphitic carbon quantum dots with size tunability and uniformity using reverse micelles.
PMID 22510781
Abstract
Highly luminescent graphitic carbon quantum dots (GQDs) are synthesized employing reverse micelles as nanoreactors. This method offers size tunability and narrow size distribution without any unpractical size separation process. Also, high quantum yields of maximum 35% at the 360 nm excitation wavelength are achieved.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
445312
Hexadecylamine, 98%
C16H35N
H7408
Hexadecylamine, technical grade, 90%
C16H35N