EMAIL THIS PAGE TO A FRIEND
Critical care medicine
Low-dose antioxidant is sufficient to regulate pulmonary redox equilibration.
PMID 22511172
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A5922
Amifostine trihydrate, ≥97% (TLC), powder
C5H15N2O3PS · 3 H2O