EMAIL THIS PAGE TO A FRIEND
Yao xue xue bao = Acta pharmaceutica Sinica
[Analysis and evaluation of the impurity of felodipine and its tablets].
PMID 22512035
Abstract
The paper reports the systematic study on felodipine and its impurities in tablets, to improve its quality standards for the control of the related substances. HPLC-DAD, UPLC-MS, IR and NMR methods were used for the isolation of felodipine and its impurities in tablets, their identification and the zebrafish animal model was used for the analysis of the toxic impurities. In felodipine material and its tablets, three impurities are isolated and identified. They are impurity 1 [dimethyl 4-(2, 3-dichlorophenyl)-2, 6-dimethyl-1, 4-dihydropyridine-3, 5-dicarboxylate], impurity 2 [ethyl methyl 4-(2, 3-dichlorophenyl)-2, 6-dimethylpyridine-3, 5-dicarboxylate] and impurity 3 [diethyl 4-(2, 3-dichlorophenyl)-2, 6-dimethyl-1, 4-dihydropyridine-3, 5-dicarboxylate], separately. The result of zebrafish animal model analysis showed that the teratogenic effects of four compounds were: impurity 3 > or = felodipine > impurity 1 > impurity 2, lethal effects were as follows: impurity 2 = impurity 3 > felodipine > or = impurity 1. This study confirmed the toxicity of three impurities in felodipine. According to the results, the paper suggested the amendments to the standard of the medicine and provided the support to the control of impurities in the manufacturing process.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
F9677
Felodipine, solid
C18H19NO4Cl2