EMAIL THIS PAGE TO A FRIEND
The journal of physical chemistry. A
Chemometric analysis of voltammetric data on metal ion binding by selenocystine.
PMID 22512274
Abstract
The behavior of selenocystine (SeCyst) alone or in the presence of various metal ions (Bi(3+), Cd(2+), Co(2+), Cu(2+), Cr(3+), Ni(2+), Pb(2+), and Zn(2+)) was studied using differential pulse voltammetry (DPV) over a wide pH range. Voltammetric data matrices were analyzed using chemometric tools recently developed for nonlinear data: pHfit and Gaussian Peak Adjustment (GPA). Under the experimental conditions tested, no evidence was found for the formation of metal complexes with Bi(3+), Cu(2+), Cr(3+), and Pb(2+). In contrast, SeCyst formed electroinactive complexes with Co(2+) and Ni(2+) and kinetically inert but electroactive complexes with Cd(2+) and Zn(2+). Titrations with Cd(2+), Co(2+), Ni(2+), and Zn(2+) produced data that were reasonably consistent with the formation of stable 1:1 M(SeCyst) complexes.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
S1650
Seleno-DL-cystine, powder
C6H12N2O4Se2
545996
Seleno-L-cystine, 95%
C6H12N2O4Se2