EMAIL THIS PAGE TO A FRIEND
Journal of the European Academy of Dermatology and Venereology : JEADV
Ocular damage in patients with psoriasis treated by psoralen UV-A therapy or narrow band UVB therapy: a systematic literature review.
PMID 22512678
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
56448
Xanthotoxin, analytical standard
C12H8O4