EMAIL THIS PAGE TO A FRIEND
International journal of cosmetic science
Lemon (Citrus limon, Burm.f.) essential oil enhances the trans-epidermal release of lipid-(A, E) and water-(B6, C) soluble vitamins from topical emulsions in reconstructed human epidermis.
PMID 22515469
Abstract
Topical bioavailability of lipid- and water-soluble vitamins is a critical issue for protecting or anti-ageing formulations. Using 17-day-old SkinEthic(®) reconstructed human epidermis, we investigated (at 34°C) the role of lemon EO in enhancing the penetration of α-tocopherol (E) and retinyl acetate (A), pyridoxine (B(6)) and ascorbic acid (C), released from O/W or W/O emulsions. D-limonene, α-pinene and p-cymene (65.9, 2.2 and 0.5%w/w of the oil) had skin permeability coefficients Ps (10(-3) cm h(-1)) of 0.56 ± 0.03 (or 0.73 ± 0.02), 0.72 ± 0.05 (or 0.98 ± 0.05) and 0.84 ± 0.04 (or 1.14 ± 0.04), respectively, when incorporated in a W/O (or O/W) emulsion. Vitamins B6, C and A had Ps values of (3.0 ± 0.4) × 10(-3), (7.9 ± 0.6) × 10(-3) and (0.37 ± 0.02) × 10(-5) cm h(-1), respectively, and their flux through the skin was enhanced by a factor of 4.1, 3.4 and 5.8, respectively, in the presence of lemon EO. The penetration of vitamin E was nine-fold enhanced. Lemon EO produced only reversible modification of TEWL, and it is a safe and effective penetration enhancer for topical administration of lipid- and water-soluble vitamins.