EMAIL THIS PAGE TO A FRIEND
Journal of colloid and interface science
Controllable synthesis of MnS nanocrystals from a single-source precursor.
PMID 22515997
Abstract
A facile single-source precursor method has been applied for the selective synthesis of MnS nanocrystals (NCs) with well-defined shapes and crystal structures such as hexapod, octahedral, hexagonal shaped α-MnS NCs, and pencil-shaped γ-MnS NCs. The effects of the composition of precursor, reaction temperature, and the heating rate on the morphologies, and crystal structures of MnS NCs were systematically studied for the first time.