EMAIL THIS PAGE TO A FRIEND
Macromolecular rapid communications
Propagation rate coefficients for homogeneous phase VDF-HFP copolymerization in supercritical CO2.
PMID 22517682
Abstract
For the first time, propagation rate coefficients, k(p,COPO) , for the copolymerizations of vinylidene fluoride and hexafluoropropene have been determined. The kinetic data was determined via pulsed-laser polymerization in conjunction with polymer analysis via size-exclusion chromatography, the PLP-SEC technique. The experiments were carried out in homogeneous phase with supercritical CO(2) as solvent for temperatures ranging from 45 to 90 °C. Absolute polymer molecular weights were calculated on the basis of experimentally determined Mark-Houwink constants. The Arrhenius parameters of k(p,COPO) vary significantly compared with ethene, which is explained by the high electronegativity of fluorine and less intra- and intermolecular interactions between the partially fluorinated macroradicals.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
295388
Hexafluoropropene, ≥99%
C3F6