EMAIL THIS PAGE TO A FRIEND
General hospital psychiatry
Risperidone-induced leukopenia: a case report and brief review of literature.
PMID 22520716
Abstract
A Caucasian, male, young adult with recurrent agitated depression and suicidal ideation received lithium and oral olanzapine. His white blood cell count was normal at that time. Due to unsatisfactory response, he received 4 mg/day risperidone. While symptoms improved, leukopenia emerged, specifically directed towards neutrophils. Upon risperidone discontinuation, white blood cell count returned to reference values within 1 week. As symptom control was satisfactory, we attempted no risperidone rechallenge. Accurate blood testing must accompany atypical antipsychotic drug administration since blood dyscrasias are always possible with these drugs.