EMAIL THIS PAGE TO A FRIEND
Chemistry & biology
Insights into quinaldic acid moiety formation in thiostrepton biosynthesis facilitating fluorinated thiopeptide generation.
PMID 22520750
Abstract
Thiostrepton (TSR), often referred as to a parent compound in the thiopeptide family, is a bimacrocyclic member that features a quinaldic acid (QA) moiety-containing side ring appended to the characteristic core system. QA biosynthesis requires an unusual ring-expanding conversion, showing a methyl transfer onto and a rearrangement of the indole part of L-tryptophan to give a quinoline ketone. Herein, we report that the process involves the activities of the radical methyltransferase TsrT, aminotransferase TsrA, dehydrogenase TsrE, and cyclase TsrD. TsrU, a stereospecific oxidoreductase, catalyzes the further conversion of the ketone into an enantiomerically pure S-alcohol. Elucidation of this chemistry, which is common in the biosynthesis of a number of thiopeptides sharing a QA side ring system, facilitates analog generation, as shown by the achievement of region-specific fluorination of thiostrepton with the improved antibacterial activity.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
160660
Quinaldic acid, 98%
C10H7NO2
T8902
Thiostrepton from Streptomyces azureus, ≥90% (HPLC)
C72H85N19O18S5