EMAIL THIS PAGE TO A FRIEND
Organic & biomolecular chemistry
Synthesis of the anti-influenza agent (-)-oseltamivir free base and (-)-methyl 3-epi-shikimate.
PMID 22522650
Abstract
A new enantioselective synthesis of the anti-influenza agent (-)-oseltamivir free base (7.1% overall yield; 98% ee) and (-)-methyl 3-epi-shikimate (16% overall yield; 98% ee) has been described from readily available raw materials. Sharpless asymmetric epoxidation and diastereoselective Barbier allylation of an aldehyde are the key reactions employed in the incorporation of chirality, while the cyclohexene carboxylic ester core was constructed through a ring closing metathesis reaction.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
S5375
Shikimic acid, ≥99%
C7H10O5
69686
Shikimic acid, analytical standard
C7H10O5