EMAIL THIS PAGE TO A FRIEND
Journal of nanoscience and nanotechnology
The influence of interlayer molecules in luminescence of self-assembled bilayer PbS quantum dot films.
PMID 22523994
Abstract
We synthesized organic solvent-soluble and water-soluble PbS quantum dots (QDs) with different sizes. The organic solvent-soluble PbS QDs dispersed in tetrachloroethylene were used to prepare bilayers structures of QDs bound by dithiol linkers on GaAs. The water-soluble PbS QDs were used to prepare bilayer structures of QDs on quartz based on alternating adsorption of polyelectrolyte. For bilayer films on GaAs, it was found that the stacking sequence of QDs affects the quantum yield and emission wavelength of the larger QDs. However, for bilayer films with different stacking sequences on quartz, the larger QDs show similar PL intensities and emission wavelength independent of the sequence. The probable mechanism for this difference observed is discussed in terms of charge transfer between QDs.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
254231
Lead(II) selenide, 99.99%
PbSe