EMAIL THIS PAGE TO A FRIEND
Clinical radiology
Re: bovine aortic arch: a novel association with thoracic aortic dilation.
PMID 22526059
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C2659
Chromomycin A3 from Streptomyces griseus, ≥95% (HPLC)
C57H82O26