EMAIL THIS PAGE TO A FRIEND
Chemical communications (Cambridge, England)
Phosphine-catalyzed intramolecular γ-umpolung addition of α-aminoalkylallenic esters: facile synthesis of 3-carbethoxy-2-alkyl-3-pyrrolines.
PMID 22526463
Abstract
An array of N-tosylated α-aminoalkylallenic esters was prepared and their cyclization under the influence of nucleophilic phosphine catalysts was explored. The α-aminoalkylallenic esters were prepared through aza-Baylis-Hillman reactions or novel DABCO-mediated decarboxylative rearrangements of allenylic carbamates. Conversion of these substrates to 3-carbethoxy-2-alkyl-3-pyrrolines was facilitated through Ph(3)P-catalyzed intramolecular γ-umpolung addition.